Learn More
EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry
Brand: Honeywell Fluka 34550-10L-VP
350.84 EUR valid until 2024-12-31
Use promo code "21986" to get your promotional price.
Code : NEW
Additional Details : Weight : 10.00000kg
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Ethylenediaminetetraacetic acid disodium salt solution | |
C10H14N2Na2O8 | |
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
3822669 | |
ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate | |
57339238 | |
336.21g/mol |
139-33-3 | |
[-CH2N(CH2CO2Na)CH2CO2H]2 | |
NONH for all modes of transport | |
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
336.21 | |
CHEBI:64734 | |
10 L |