missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Solstice

Product Code. 15644280
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
10 L
6 x 1 L
Unit Size:
1 L
10 L
2.5 L
5 L
500mL
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15644280 6 x 1 L Pack of 6
15614280 500 mL 500mL
15694270 1 L 1 L
15604280 2.5 L 2.5 L
15654280 6 x 500 mL Pack of 6
15635735 5 L 5 L
15624280 5 L 5 L
15625735 10 L 10 L
15674270 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15644280 Supplier Honeywell-Fluka Supplier No. 345506X1L
  • ONLINE EXCLUSIVE PRICE
    353.00 €
    115.33 € / Pack of 6
    Save 237.67 €
    67% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA disodium salt solution
Name Note Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry
CAS 139-33-3, 7732-18-5
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Molecular Formula C10H14N2Na2O8
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Beilstein 3822669
Concentration 0.1 M EDTA-Na2
Density 1.01 g/cm3
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Formula Weight 372.24
Grade Reagent Ph. Eur.
Packaging Poly Bottle
Quantity 6 x 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.