missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Efaproxiral sodium, 98%

Product Code. 15556006 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15556006 250 mg 250mg
15546006 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15556006 Supplier Thermo Scientific Chemicals Supplier No. 460102500
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 170787-99-2 | C20H22NNaO4 | 363.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170787-99-2
Molecular Formula C20H22NNaO4
Molecular Weight (g/mol) 363.38
InChI Key SWDPIHPGORBMFR-UHFFFAOYSA-M
Synonym efaproxiral sodium, sodium 2-4-2-3,5-dimethylphenyl amino-2-oxoethyl phenoxy-2-methylpropanoate, rsr 13 sodium, unii-3l83qp52xi, rsr13 sodium, efaproxiral sodium usan, rsr-13
PubChem CID 2725048
IUPAC Name sodium;2-[4-[2-(3,5-dimethylanilino)-2-oxoethyl]phenoxy]-2-methylpropanoate
SMILES CC1=CC(=CC(=C1)NC(=O)CC2=CC=C(C=C2)OC(C)(C)C(=O)[O-])C.[Na+]

Specifications

Loss on Drying 13% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 363.38
Percent Purity 98%
Chemical Name or Material Efaproxiral sodium
compliance-icons
Product Identifier
  • Efaproxiral sodium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.