missing translation for 'onlineSavingsMsg'
Learn More

Efavirenz Amino Alcohol Ethyl Carbamate, TRC

Product Code. 30179599
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179599 10 mg 10mg
30179471 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179599 Supplier Toronto Research Chemicals Supplier No. TRCE42501510MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211563-41-6
Molecular Formula C16 H15 Cl F3 N O3
Molecular Weight (g/mol) 361.74
Synonym Carbamic acid, [4-chloro-2-[(1S)-3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propynyl]phenyl]-, ethyl ester (9CI), [4-Chloro-2-[(1S)-3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propynyl]phenyl]carbamic Acid Ethyl Ester, Ethyl N-[4-Chloro-2-[(1S)-3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)prop-2-yn-1-yl]phenyl]carbamate
IUPAC Name ethyl N-[4-chloro-2-[(2S)-4-cyclopropyl-1,1,1-trifluoro-2-hydroxybut-3-yn-2-yl]phenyl]carbamate
SMILES CCOC(=O)Nc1ccc(Cl)cc1[C@@](O)(C#CC2CC2)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H15ClF3NO3/c1-2-24-14(22)21-13-6-5-11(17)9-12(13)15(23,16(18,19)20)8-7-10-3-4-10/h5-6,9-10,23H,2-4H2,1H3,(H,21,22)/t15-/m0/s1
Formula Weight 361.0693
Chemical Name or Material Efavirenz Amino Alcohol Ethyl Carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.