missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Efavirenz, MedChemExpress
Description
Efavirenz (DMP 266) is a potent inhibitor of the wild-type HIV-1 reverse transcriptase with a Ki of 2.93 nM and exhibits an IC95 of 1.5 nM for the inhibition of HIV-1 replicative spread in cell culture.
Specifications
Specifications
| Chemical Name or Material | Efavirenz |
| Physical Form | Powder |
| CAS | 154598-52-4 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H9ClF3NO2 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | DMP 266 EFV L-743726 |
| Solubility Information | DMSO : ≥ 38 mg/mL (120.38 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?