missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCBP2 Antibody (CL0524) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52951JF669
This item is not returnable.
View return policy
Description
EFCBP2 Monoclonal antibody specifically detects EFCBP2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| EFCBP2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EF-hand calcium binding protein 2, N-terminal EF-hand calcium binding protein 2 | |
| This antibody was developed using a recombinant protein derived from Q7Z6G3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL0524 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 54550 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction