Learn More
EFCBP2 Antibody (CL0524) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52951MFV500
Product Code. 30296630
  • 759.00 € / 0.10mL
Description
EFCBP2 Monoclonal antibody specifically detects EFCBP2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EFCBP2
Monoclonal
mFluor Violet 500 SE
EF-hand calcium binding protein 2, N-terminal EF-hand calcium binding protein 2
This antibody was developed using a recombinant protein derived from Q7Z6G3, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL0524
50mM Sodium Borate
Mouse
Protein A purified
RUO
54550
Store at 4⪚C in the dark.
IgG1