missing translation for 'onlineSavingsMsg'
Learn More

Efinaconazole, MedChemExpress

Product Code. 16482807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482807 50 mg 50mg
16479277 5 mg 5mg
16466866 100 mg 100mg
16466728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482807 Supplier Medchem Express Supplier No. HY15660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Efinaconazole (KP-103) is a triazole antifungal agent and againsts T. mentagrophytes SM-110 and C. albicans ATCC 10231 with MICs of 0.0039 μg/mL and 0.00098 μg/mL, respectively. Efinaconazole has a potent in vitro activity against fungal pathogens including dermatophytes, Candida and Malassezia species.

Efinaconazole (KP-103) is a triazole antifungal agent and againsts T. mentagrophytes SM-110 and C. albicans ATCC 10231 with MICs of 0.0039 μg/mL and 0.00098 μg/mL, respectively. Efinaconazole has a potent in vitro activity against fungal pathogens including dermatophytes, Candida and Malassezia species.

Specifications

Chemical Name or Material Efinaconazole
Physical Form Solid
CAS 164650-44-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H22F2N4O
For Use With (Application) COVID-19-immunoregulation
Synonym KP-103
Solubility Information DMSO : ≥ 100 mg/mL (287.03 mM)
SMILES C[C@@H](N1CCC(CC1)=C)[C@@](O)(C2=CC=C(F)C=C2F)CN3N=CN=C3
Molecular Weight (g/mol) 348.39
Formula Weight 348.39
Percent Purity 99.36%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.