missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eGFP Monoclonal Antibody (F56-6A1.2.3)
Description
MA1-952 contains 100 μg of in vitro produced, Protein A purified IgG (1 mg/mL) in PBS containing 1 mg/mL BSA and 0.05% sodium azide. MA1-952 detects eGFP and eGFP tagged proteins. MA1-952 has been successfully used in Western blot and immunofluorescence applications. The MA1-952 immunogen is full length enhanced green fluorescent protein (eGFP).
eGFP (green fluorescent protein) fluorescent protein.
Specifications
Specifications
| Antigen | eGFP |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F56-6A1.2.3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene Alias | eGFP; egfp tag; GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Green Fluoresecent Protein; Turbo eGFP |
| Host Species | Mouse |
| Immunogen | Full length enhanced green fluorescent protein (eGFP). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?