missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Antibody (B1D8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
595.00 € - 642.00 €
Specifications
| Antigen | EGFR |
|---|---|
| Clone | B1D8 |
| Dilution | Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunoaffinity Purification, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Affinity Purification, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18404752
|
Novus Biologicals
NBP2-34553B-100ul |
100 μL |
642.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18401632
|
Novus Biologicals
NBP2-34553B |
0.1 mL |
595.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGFR Monoclonal antibody specifically detects EGFR in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Affinity Purification, CyTOFSpecifications
| EGFR | |
| Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunoaffinity Purification, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse (Negative), Rat (Negative) | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Microsomes from A431 cells. | |
| Primary | |
| Store at 4C in the dark. |
| B1D8 | |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Affinity Purification, CyTOF | |
| Biotin | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
| PBS | |
| 1956 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title