missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Antibody (EGFR/8933R), Novus Biologicals™
Rabbit Monoclonal Antibody
291.00 € - 530.00 €
Specifications
| Antigen | EGFR |
|---|---|
| Clone | EGFR/8933R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613655
|
Novus Biologicals
NBP3-23766-20ug |
20 μg |
291.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631795
|
Novus Biologicals
NBP3-23766-100ug |
100 μg |
530.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| EGFR | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
| 10 mM PBS with 0.05% BSA | |
| 1956 | |
| IgG κ | |
| Protein A or G purified |
| EGFR/8933R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Recombinant fragment (around aa300-500) of human EGFR protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title