missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Mouse, Clone: 31G7, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1956-MSM4-P1
Additional Details : Weight : 0.01000kg
Specifications
EGFR | |
Cell surface | |
31G7 | |
Unconjugated | |
NP_001333828 | |
Mouse | |
∽170kDa (wild type), ∽145kDa (vIII variant) | |
Protein A/G Purified | |
RUO | |
1956 | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide | |
EGFR | |
Human EGFR purified from A431 cells | |
A431 cells. Placenta, Breast, Colon or Bladder cancer | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |