missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Mouse, Clone: GFR/1667, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1956-MSM7-P0
This item is not returnable.
View return policy
Specifications
| EGFR | |
| Cell surface | |
| GFR/1667 | |
| Unconjugated | |
| NP_001333828 | |
| Mouse | |
| 170kDa (wild type) and 145kDa (vIII variant) | |
| Protein A/G Purified | |
| RUO | |
| 1956 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| EGFR | |
| Recombinant human full-length EGFR protein | |
| A431 cells. Placenta, Breast, Colon or Bladder cancer | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction