Learn More
EGLN3/PHD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 46 publications
Brand: Novus Biologicals NB100-139
This item is not returnable.
View return policy
Description
EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy, Mass Spectrometry, Knockdown Validated.
Specifications
| EGLN3/PHD3 | |
| Polyclonal | |
| EGLN3 | |
| Affinity Purified | |
| IgG |
| Immunohistochemistry, Immunoassay, KnockDown | |
| EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
| 28 kDa | |
| Mouse, Rat |