missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EGR1 Monoclonal Antibody (406913)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561207

Product Code. 30533678

  • 499.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EGR1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P18146
EGR1
E. coli-derived recombinant human EGR1, Gln282-Ser433.
100 μg
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin)
406913
Unconjugated
EGR1
A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225
Rat
Protein A/G
RUO
1958
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.