missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92512-0.1ml
This item is not returnable.
View return policy
Description
EGR3 Polyclonal antibody specifically detects EGR3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| EGR3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| early growth response 3, early growth response protein 3, EGR-3, MGC138484, PILOTZinc finger protein pilot | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human EGR3 (NP_004421.2). MTGKLAEKLPVTMSSLLNQLPDNLYPEEIPSALNLFSGSSDSVVHYNQMATENVMDIGLTNEKPNPELSYSGSFQPAPGNKTVTYLGKFAFDSPSNWCQDNIISLMSAGILGVPPASGALSTQTSTASMVQPPQGDVEAMYPALPPYSNCGDLYSEPVSFHDPQGNPGLAYSPQDYQSAKPALDSNLFPMIPDYNLYHHPNDMGSIPEHKPFQGMDPIRVNPPPITPLETIKAFKDKQIHPGFGSLPQPP | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 1960 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction