missing translation for 'onlineSavingsMsg'
Learn More

EGTA, TRC

Product Code. 30012758
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012758 100 g 100g
30011043 1 g 1g
30003907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012758 Supplier Toronto Research Chemicals Supplier No. TRCE440150100G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14 H24 N2 O10
Molecular Weight (g/mol) 380.35
Synonym EGTA, 2,2'-Ethylenedioxybis(ethylamine)-N,N,N',N'-tetraacetic Acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic Acid, 3,6-Dioxaoctane-1,8-diamine-N,N,N',N'-tetraacetic Acid, Chelest GEA, EBONTA, EDGA, EGTA, Egtazic Acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N',N'-tetraacetic Acid, Ethylene Glycol Bis(β-aminoethyl Ether)-N,N,N',N'-tetraacetic Acid, GEDTA, NSC 615010, O,O'-Bis(2-aminoethyl)ethylene Glycol-N,N,N',N'-tetraacetic Acid, [Ethylenebis(oxyethylenenitrilo)]tetraacetic Acid, [Ethylenebis(oxyethylenenitrilo)]tetraacetic Acid
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCOCCOCCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C14H24N2O10/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)
Formula Weight 380.1431
Chemical Name or Material 1,2-Bis(2-aminoethoxyethane)-N,N,N',N'-tetraacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.