missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHF Recombinant Rabbit Monoclonal Antibody (7O0N9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558134
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: MILEGGGVMN LNPGNNLLHQ PPAWTDSYST CNVSSGFFGG QWHEIHPQYW TKYQVWEWLQ HLLDTNQLDA NCIPFQEFDI NGEHLCSMSL QEFTRAAGTA GQLLYSNLQH LKW.
This gene encodes a protein that belongs to an ETS transcription factor subfamily characterized by epithelial-specific expression (ESEs). The encoded protein acts as a transcriptional repressor and may be associated with asthma susceptibility. This protein may be involved in epithelial differentiation and carcinogenesis.
Tekniske data
| EHF | |
| Recombinant Monoclonal | |
| 1.28 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9NZC4 | |
| EHF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human EHF (NP_036285.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunoprecipitation | |
| 7O0N9 | |
| Unconjugated | |
| EHF | |
| 9030625L19Rik; AU019492; EHF; Epithelium-specific Ets transcription factor 3; ESE3; ESE-3; ESE3 transcription factor; ESE3B; ESEJ; ETS domain-containing transcription factor; ETS homologous factor; hEHF; Unknown (protein for MGC:133971) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 26298 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion