missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic acid ethyl ester, MedChemExpress

Product Code. 16492206
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492206 500 mg 500mg
16484467 50 mg 50mg
16418446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492206 Supplier Medchem Express Supplier No. HYB0747
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Eicosapentaenoic acid ethyl ester is an omega-3 fatty acid agent.

Eicosapentaenoic acid ethyl ester is an omega-3 fatty acid agent.

Specifications

Chemical Name or Material Eicosapentaenoic acid ethyl ester
Physical Form Liquid
CAS 86227-47-6
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C22H34O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym EPA ethyl ester Ethyl eicosapentaenoate
Solubility Information Ethanol : 50 mg/mL (151.29 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (100.85 mM; Need ultrasonic)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(OCC)=O
Molecular Weight (g/mol) 330.5
Formula Weight 330.5
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.