missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic Acid Ethyl Ester, TRC

Product Code. 30180191
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180191 10 mg 10mg
30180078 50 mg 50mg
30179743 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180191 Supplier Toronto Research Chemicals Supplier No. TRCE47780510MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86227-47-6
Molecular Formula C22 H34 O2
Molecular Weight (g/mol) 330.5
Synonym (5Z,8Z,11Z,14Z,17Z)-5,8,11,14,17-Eicosapentaenoic acid ethyl ester, 5,8,11,14,17-Eicosapentaenoic acid, ethyl ester, (all-Z)-, (all-Z)-5,8,11,14,17-Eicosapentaenoic acid ethyl ester, AMR 101, Epadel, Epadel S 300, Ethyl all-cis-5,8,11,14,17-eicosapentaenoate, Ethyl eicosapentaenoate, Ethyl eicosapentanoate, Icosapent ethyl, Incromega EPA, Timnodonic acid ethyl ester, Vascepa, cis-Eicosapentaenoic acid ethyl ester
IUPAC Name ethyl (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoate
SMILES CCOC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-4-2/h5-6,8-9,11-12,14-15,17-18H,3-4,7,10,13,16,19-21H2,1-2H3/b6-5-,9-8-,12-11-,15-14-,18-17-
Formula Weight 330.2559
Chemical Name or Material Eicosapentaenoic Acid Ethyl Ester (EPA EE)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.