missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84876
This item is not returnable.
View return policy
Description
EIF3A Polyclonal antibody specifically detects EIF3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EIF3A | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| centrosomin homolog, cytoplasmic protein p167, eIF3 p167, eIF3 p180, eIF3 p185, EIF3, p180 subunit, eIF3a, eIF3-p170, EIF3S10EIF3, eIF-3-theta, eIF3-theta, Eukaryotic translation initiation factor 3 subunit 10, eukaryotic translation initiation factor 3 subunit A, eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD), eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD), eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa, eukaryotic translation initiation factor 3, subunit 10, 170kD, eukaryotic translation initiation factor 3, subunit A, KIAA0139P167, p180, p185, TIF32 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LQNNTILRLLQQVSQIYQSIEFSRLTSLVPFVDAFQLERAIVDAARHCDLQVRIDHTSRTLSFGSDLNYATREDAPIG | |
| 0.1 mL | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 8661 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction