Learn More
Elafin/Skalp Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15963-1-AP-150UL
 View more versions of this product
Product Code. 16865374
  • 410.00 € / 150µL
Description
Neutrophil and pancreatic elastase-specific inhibitor of skin. It may prevent elastase-mediated tissue proteolysis.
Specifications
Specifications
Elafin/Skalp
Polyclonal
Unconjugated
PI3
cementoin, Elafin, Elastase specific inhibitor, ESI, Peptidase inhibitor 3, PI 3, PI3, Protease inhibitor WAP3, SKALP, WAP3, WFDC14
Rabbit
Antigen Affinity Chromatography
RUO
5266
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P19957
PI3
Elafin/Skalp Fusion Protein Ag8735
150 μL