missing translation for 'onlineSavingsMsg'
Learn More

Elaidic acid, MedChemExpress

Product Code. 16415566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16415566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16415566 Supplier Medchem Express Supplier No. HY113016
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Elaidic acid is the major trans fat found in hydrogenated vegetable oils and can be used as a pharmaceutical solvent.

Elaidic acid is the major trans fat found in hydrogenated vegetable oils and can be used as a pharmaceutical solvent.

Specifications

Chemical Name or Material Elaidic acid
Physical Form Oil
CAS 112-79-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H34O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 13.5 mg/mL (47.79 mM; Need ultrasonic)
SMILES CCCCCCCC/C=C/CCCCCCCC(O)=O
Molecular Weight (g/mol) 282.46
Formula Weight 282.46
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.