missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Elastin Recombinant Rabbit Monoclonal Antibody (9F5R1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558058
This item is not returnable.
View return policy
Description
Immunogen sequence: GGVPGAIPGG VPGGVFYPGA GLGALGGGAL GPGGKPLKPV PGGLAGAGLG AGLGAFPAVT FPGALVPGGV ADAAAAYKAA KAGAGLGGVP GVGGLGVSAG AVVPQPGAGV KPGKVPGVGL PGVYPGGVLP GARFPGVGVL PGVPTGAGVK PKAPGVGGAF AGIPGVGPFG GPQP.
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| Elastin | |
| Recombinant Monoclonal | |
| 0.41 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P15502 | |
| ELN | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9F5R1 | |
| Unconjugated | |
| ELN | |
| 5830419M17Rik; AI385707; AI480567; AW229038; BB105748; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; Eln; EMI; EMILIN; EMILIN1; EMILIN-1; FLJ38671; FLJ43523; gp115; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2006 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction