missing translation for 'onlineSavingsMsg'
Learn More

Elesclomol, MedChemExpress

Product Code. 16491187
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491187 50 mg 50mg
16437687 5 mg 5mg
16424948 10 mg 10mg
16405726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491187 Supplier Medchem Express Supplier No. HY12040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elesclomol (STA-4783) is a potent copper ionophore and promotes copper-dependent cell death (cuproptosis).

Elesclomol specifically binds ferredoxin 1 (FDX1) α2/α3 helices and β5 strand. Elesclomol inhibits FDX1-mediated Fe-S cluster biosynthesis. Elesclomol is an oxidative stress inducer that induces cancer cell apoptosis. Elesclomol is a reactive oxygen species (ROS) inducer. Elesclomol can be used for Menkes and associated disorders of hereditary copper deficiency research.

Specifications

Chemical Name or Material Elesclomol
Physical Form Solid
CAS 488832-69-5
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H20N4O2S2
For Use With (Application) Cancer-programmed cell death
Synonym STA-4783
Solubility Information DMSO : ≥ 22 mg/mL (54.93 mM)
SMILES S=C(C1=CC=CC=C1)N(NC(CC(NN(C(C2=CC=CC=C2)=S)C)=O)=O)C
Molecular Weight (g/mol) 400.52
Formula Weight 400.52
Percent Purity 98.46%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.