missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF4 Monoclonal Antibody (OTI1E4), TrueMAB™, OriGene
Description
ELF4 Monoclonal antibody specifically detects ELF4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ELF4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1E4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Elf4 |
| Gene Accession No. | Q99607 |
| Gene Alias | AV314029; BC042423; E74 like ETS transcription factor 4; E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ELFR; ENSMUSG00000053512; ETS-related transcription factor Elf-4; Gm9907; MEF; myeloid elf-1 like factor; myeloid Elf-1-like factor; RGD1560743 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?