Learn More
GVS SPR503 Elipse Mask with A1P3 Filters
Click to view available options
:
GVS SPR503 Elipse Mask
Description
This mask is recommended for organic gases and dust, pesticides, painting, solvents into automotive, construction, agriculture and shipyard industry or repair.
- Extremely lightweight and compact to give the wearer as much comfort as possible. It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort. Filters last up to 3 years and can be easily replaced.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
- Certifications: EN140: 1998 Mask Body. A1P3 R D Filters EN14387: 2004 + A1: 2008 (suited for organic gases and dust). CE 2797
Specifications
Specifications
| Color | Blue |
| Disposable | Reusable |