missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELK1(phospho T417) & ELK1 Protein Phosphorylation Antibody Pair, Abnova™
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the ELK1 protein, and the other against the specific T417 phosphorylated site of ELK1 for use in in situ Proximity Ligation Assay.
Specifications
Specifications
| Antigen | ELK1 |
| Applications | In situ PLA |
| Conjugate | Unconjugated |
| Gene | ELK1, member of ETS oncogene family |
| Gene Alias | - |
| Gene Symbols | ELK1 |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 2002 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?