missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 539.00 €
Specifications
| Antigen | ELK3 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) -Reported in scientific literature (PMID: 25401928) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), ChIP Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18466800
|
Novus Biologicals
NBP1-83960-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18436510
|
Novus Biologicals
NBP1-83960 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELK3 Polyclonal antibody specifically detects ELK3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP)Specifications
| ELK3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ELK3, ETS-domain protein (SRF accessory protein 2), ERPETS domain-containing protein Elk-3, ETS-domain protein, ETS-related protein ERP, NETSRF accessory protein 2, SAP-2, SAP2ETS-related protein NET, Serum response factor accessory protein 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KNIIKKVIGQKFVYKFVSFPEILKMDPHAVEISRESLLLQDSDCKASPEGREAHKHGLAALRSTSRNEYIHSGLYSSFTINSLQNPPDAFKAIKTEKLEEPPEDSPPVEEVRTVIRFVTNKTDKHVTR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Chromatin Immunoprecipitation (ChIP) -Reported in scientific literature (PMID: 25401928) | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 2004 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title