missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ellagic acid, 97%

Product Code. 10550794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10550794 2.5 g Glass bottle 2.5g
10695722 10 g Glass bottle 10g
10464375 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10550794 Supplier Thermo Scientific Chemicals Supplier No. 117740025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 476-66-4 | C14H6O8 | 302.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Melting Point >360°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 2.5 g
Beilstein 19, 261
Merck Index 15, 3602
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkalies and pyridine,practically insoluble in ether
Formula Weight 302.19
Percent Purity 97%
Chemical Name or Material Ellagic acid
compliance-icons
Product Identifier
  • Ellagic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.