missing translation for 'onlineSavingsMsg'
Learn More

Ellagic acid hydrate, 97%, may cont. up to 12% water

Product Code. 11492348
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492348 100 g 100g
11472348 5 g 5g
11482348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492348 Supplier Thermo Scientific Alfa Aesar Supplier No. A15722.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 476-66-4 | C14H6O8 | 302.19 g/mol

Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.

Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Melting Point 450°C (decomposition)
Quantity 100 g
Beilstein 47549
Sensitivity Air and light sensitive
Merck Index 14,3547
Solubility Information Slightly soluble in water. Soluble in 1M sodium hydroxide (10mg/ml),EtOH,and pyridine.
Formula Weight 302.20 (Anhydrous)
Percent Purity 97%
Assay may cont. up to 12% water
Chemical Name or Material Ellagic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.