missing translation for 'onlineSavingsMsg'
Learn More

Ellagic acid, MP Biomedicals™

Product Code. 11496832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496832 1 g 1g
11416842 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496832 Supplier MP Biomedicals Supplier No. 0215102901
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Color Brown-Yellow
Quantity 1 g
Formula Weight 302.2
Percent Purity 0.999
Physical Form Powder
Chemical Name or Material Ellagic acid, For GC analysis
compliance-icons
Product Identifier
  • Ellagic Acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ellagic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.