missing translation for 'onlineSavingsMsg'
Learn More

Ellagic Acid, MP Biomedicals™

Product Code. 11406842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406842 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406842 Supplier MP Biomedicals Supplier No. 0215102905
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the assay of factor XIIa in plasma. Ellagic Acid, MP Biomedicals is a commonly occurring plant polyphenol and an inhibitor of glutathione S-transferase with contact activation in blood coagulation.

Ellagic acid is a naturally occurring plant phenol that is a potent antioxidant, presumably by acting as a scavenger of oxygen species produced by hydrogen peroxide treatment. It acts as a protector of the DNA double helix from alkylating agent injury.

  • Delayed cutaneous hypersensitivity reaction to tuberculin and bovine serum albumin in guinea pigs
  • Has anti-mutagenic and anti-carcinogenic properties
  • Reported to inhibit topoisomerases I and II and gyrase
  • Inhibitor of glutathione S-transferase
  • Hemostatic agent

Specifications

CAS 476-66-4
Melting Point >300°C (literature)
Molecular Formula C14H6O8
MDL Number MFCD00006914
Quantity 5 g
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
Solubility Information Soluble in 1M Sodium Hydroxide (10mg/mL, dark brown to green solution) or other alkalies; soluble in pyridine. Slightly soluble in water or ethanol.
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
Molecular Weight (g/mol) 302.19
PubChem CID 5281855
ChEBI CHEBI:4775
Formula Weight 302.2
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.