missing translation for 'onlineSavingsMsg'
Learn More

Ellagic Acid, TRC

Product Code. 30012014
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012014 500 mg 500mg
30012792 1 g 1g
17973525 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012014 Supplier Toronto Research Chemicals Supplier No. TRCE505050500MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14 H6 O8
Molecular Weight (g/mol) 302.19
Synonym 2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione, 2,3,7,8-Tetrahydroxy-chromeno[5,4,3-cde]chromene-5,10-dione, Alizarin yellow, Alizarine Yellow, Benzoaric acid, C.I. 55005, C.I. 75270, Elagostasine, Eleagic acid, Ellagic acid, Ellagic acid LT, G 91006, Gallogen, Gallogen (astringent), LDN 0097519, Lagistase, [1,1'-Biphenyl]-2,2'-dicarboxylic acid, 4,4',5,5',6,6'-hexahydroxy-, di-δ-lactone
SMILES Oc1cc2C(=O)Oc3c(O)c(O)cc4C(=O)Oc(c1O)c2c34

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H6O8/c15-5-1-3-7-8-4(14(20)22-11(7)9(5)17)2-6(16)10(18)12(8)21-13(3)19/h1-2,15-18H
Formula Weight 302.0063
Percent Purity >95%
Chemical Name or Material Ellagic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.