missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELMOD3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15913755
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15913755 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15913755 Supplier Invitrogen™ Supplier No. PA577242

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RBM29 (RNA binding motif protein 29), also known as ELMOD3 (ELMO domain-containing protein 3) or RBED1 (RNA-binding motif and ELMO domain-containing protein 1), is a 381 amino acid protein that contains one ELMO domain and exists as six alternatively spliced isoforms. The gene encoding RBM29 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMOD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ELMOD3
Gene Accession No. Q5XIQ2, Q91YP6, Q96FG2
Gene Alias AI844780; C330008I15Rik; deafness, autosomal recessive 88; DFNB88; ELMO domain containing 3; ELMO domain-containing protein 3; ELMO/CED-12 domain containing 3; Elmod3; Kcmf1; liver-specific organic anion transporter 3TM12; LST3; organic anion transporter LST-3b; potassium channel modulatory factor 1; PP4068; Rbed1; RBM29; RNA binding motif and ELMO domain 1; RNA binding motif and ELMO/CED-12 domain 1; RNA binding motif protein 29; RNA-binding motif and ELMO domain-containing protein 1; RNA-binding motif protein 29; RNA-binding protein 29
Gene Symbols ELMOD3
Host Species Rabbit
Immunogen Recombinant full length Human ELMOD3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232089, 297342, 84173
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.