missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elongin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87040
This item is not returnable.
View return policy
Description
Elongin A Polyclonal antibody specifically detects Elongin A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Elongin A | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EloA, Elongin 110 kDa subunit, elongin-A, FLJ38760, FLJ42849, RNA polymerase II transcription factor SIII subunit A1, SIII, SIII p110, TCEB3A, transcription elongation factor B (SIII), polypeptide 3 (110kD, elongin A), transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A), transcription elongation factor B alpha subunit, transcription elongation factor B polypeptide 3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ERHLGEPHGKGVVSQNKEHKSSHKDKRPVDAKSDEKASVVSREKSHKALSKEENRRPPSGDNAREKPPSSGVKKEKDREGSSLKKKCLPPSEAASDNHLKKPKHRDPEKAKLDKSKQGLDSFDTGKGAGD | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 6924 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction