missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL5 Antibody (Z88) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00 € - 486.00 €
Specifications
| Antigen | ELOVL5 |
|---|---|
| Clone | Z88 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1, ELISA 1:1, Immunohistochemistry 1:1 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18187709
|
Novus Biologicals
NBP2-54809 |
0.1 mg |
486.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682649
|
Novus Biologicals
NBP2-54809-0.025mg |
0.025 mg |
190.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELOVL5 Monoclonal specifically detects ELOVL5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| ELOVL5 | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human | |
| dJ483K16.1, EC 2.3.1.n8, elongation of very long chain fatty acids protein 5, ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, ELOVL2,3-keto acyl-CoA synthase ELOVL5, Fatty acid elongase 1, hELO1, HELO1homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2, homolog of yeast long chain polyunsaturated fatty acid elongatio, SUR4/Elo3-like, yeast) | |
| ELOVL5 | |
| IgG | |
| Protein A purified |
| Z88 | |
| Western Blot 1:1, ELISA 1:1, Immunohistochemistry 1:1 | |
| Monoclonal | |
| Mouse | |
| Cardiovascular Biology, Cellular Signaling | |
| PBS with 0.02% Sodium Azide | |
| 60481 | |
| Peptide Sequence NNVKPRKLR | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title