Learn More
Description
The only plates with a simple “plug and play” protocol for scaffold-free, self-assembly spheroid formation at large volumes.
Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters. The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well. Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats.
•Spheroids may be formed and cultured for 21 or more days
•Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
•The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
•Black opaque sidewalls to reduce well-to-well “cross-talk”
•Well suited for luorescent/luminescent assays
•Available in multiple formats, two well geometries and surface coating options.
Specifications
Specifications
| Barcode | No Barcode |
| Breakapart | No |
| Lid | With Lid |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Binding Type | Plasma Treated |
| Color | Black, Clear Bottom |
| Product Type | Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.