missing translation for 'onlineSavingsMsg'
Learn More

Eluent 3: 0.5M Na2CO3

Product Code. 30320762
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 30320762

Brand: VHG Labs VHGIELUENT3500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium carbonate
CAS 497-19-8
Analyte or Component Names Sodium carbonate
Shipping Condition Room Temperature
Molecular Formula C O3 . 2 Na
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Carbonic Acid Disodium Salt, Bisodium Carbonate, Calcined Soda, Carbonic Acid Sodium Salt, Dense Ash, V 20 N, V 20 N, V 20 N, V 20 N, V 20 N
InChI Formula InChI=1 S/CH2O3.2 Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
Solution Type Single Solution
SMILES [Na+].[Na+].[O-]C(=O)[O-]
IUPAC Name disodium;carbonate
Solvent or Matrix Water
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 105.988
Quantity 500 mL
Formula Weight 105.964
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.