Learn More
Tuttnauer ELV-D Vertical Autoclave
The ELV-D Vertical Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
13824.36 € - 35488.11 €
Specifications
Product Type | Autoclave, Top-loading |
---|---|
Temperature (English) | 221°F to 280°F |
Sterility | Non-sterile |
Product Code | Brand | Model | Capacity (Metric) | Features | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Capacity (Metric) | Features | Price | Quantity & Availability | |||
17801077
|
Tuttnauer
2840ELV-WR-230-D |
2840ELVWR-D | 31 L | Water Reservoir |
13824.36 €
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||
17871087
|
Tuttnauer
3850ELV-WR-230-1PH-D |
3850ELVWR-D | 65 L | Single Phase, Water Reservoir |
18844.57 €
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||
17811077
|
Tuttnauer
3850ELV-380-D |
3850ELV-D | 65 L | Three Phase |
18589.23 €
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||
17801097
|
Tuttnauer
3870ELV-WR-230-D |
3870ELV-D | 85 L | Single Phase 230V, Water Reservoir |
20346.59 €
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||
17891087
|
Tuttnauer
3870ELV-WR-230-1PH-D |
3870ELV-WR-230-1PH-D | 85 L | Single Phase 230V, Water Reservoir | N/A | ||||
17818998
|
Tuttnauer
5075ELVP-WR-230-D |
5075ELV-D | 160 L | Single Phase 230V, Water Reservoir |
27300.95 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17878988
|
Tuttnauer
5075ELVCPVGP-230-D |
5075ELV-D | 160 L | Single Phase, Cooling, Vacuum, Steam Generator, Printer |
33226.99 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17868988
|
Tuttnauer
5075ELVP-WR-380-D |
5075ELV-D | 160 L | Three Phase, Water Reservoir |
27300.95 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17808998
|
Tuttnauer
5075ELVCVPGP-380-D |
5075ELV-D | 160 L | Three Phase, Cooling, Vacuum, Steam Generator, Printer |
33226.99 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17828998
|
Tuttnauer
5075ELVCPVGBHP-380-D |
5075ELV-D | 160 L | Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer |
35488.11 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Designed for your laboratory applications. The 5075ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
- Advanced Control System - Take Advantage of Tuttnauer's state-of-the-art control system with multi-color display
- Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
- Optional Internal Printing - Documents date, time, temperature and pressure
- Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
- High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
- Diverse Load Types - The 5075ELV-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
Autoclave, Top-loading | |
221°F to 280°F | |
Non-sterile |