Learn More
Invitrogen™ Silencer™ Human GPCR siRNA Library
Product Code. 15611852
Change view
Click to view available options
Quantity:
15 Plates
Unit Size:
1 set
Product Code. Quantity Unit Size
15611852 15 Plates 1 set
1 options
Product Code. 15611852 Supplier Invitrogen™ Supplier No. A30082
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Silencer™ Human GPCR siRNA Library targets 1,134 Invitrogen™ Silencer siRNAs representing 378 non-olfactory human GPCRs.
G-protein–coupled receptors (GPCRs) are key components of many signal transduction pathways and also represent an important class of druggable genes. The Silencer™ Human GPCR siRNA Library targets 1,134 Ambion™ Silencer siRNAs representing 378 non-olfactory human GPCRs. This library is designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC).
Why use the Silencer Human GPCR siRNA Library?Together with careful bioinformatics selection of gene targets, Silencer siRNAs are optimized for design and high quality synthesis with guaranteed silencing. The Silencer Human GPCR siRNA Library contains our pre-designed and when available, validated siRNAs generated with maximum potency and specificity in mind so you can focus on data quality. All Silencer libraries are selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.Silencer siRNA libraries allow you to screen efficiently and maximize data outputInvitrogen Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps to ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human GPCR siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).
Utilizing Invitrogen Silencer technology built on stringent bioinformatics to enable high target specificity and siRNA potency, the Silencer Human GPCR siRNA Library allows accurate yet cost-effective RNAi functional screening against known GPCRs.
  • Comprehensive—Expedite functional gene discovery with pre-defined Silencer Human GPCR siRNA Library
  • Reliable—Increase target screening accuracy with three individual siRNAs per gene target
  • Effective—Reduce off target impacts at low concentrations with proven Invitrogen Silencer siRNA design
  • Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats
TRUSTED_SUSTAINABILITY
Specifications
Description 1134 total siRNAs targeting 378 genes plated onto 96-well plates (15 plates)
Quantity 15 Plates
For research use only. Not for use in diagnostic procedures