Learn More
Invitrogen™ Silencer™ Select Human Cancer Genome siRNA Library
Artikelnummer. 15641872
Ansicht ändern
Click to view available options
Silencer™ Select Human Cancer Genome siRNA Library
Artikelnummer.
15641872 Silencer™ Select Human Cancer Genome siRNA Library
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15641872 Lieferant Invitrogen™ Lieferanten-Nr. A30142
 Restricted Item
Estimated Shipment: 18-02-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The Silencer™ Select Human Cancer Genome siRNA Library contains 1596 Invitrogen™ Silencer Select siRNAs targeting 532 gene targets that are commonly affected in cancers.
The Silencer™ Select Human Cancer Genome siRNA Library contains 1596 Ambion™ Silencer Select siRNAs targeting 532 gene targets that are commonly affected in cancers. This library of highly potent, chemically-modified siRNAs was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC) as well as The Cancer Genome Atlas (TCGA).
Why use the Silencer Select Human Cancer Genome siRNA Library?Together with careful bioinformatics selection of gene targets, Silencer Select siRNAs are optimized with latest design algorithms, proprietary chemical modifications and high quality synthesis to help ensure desired RNAi outcomes. The Silencer Select Human Cancer Genome siRNA Library contains our pre-designed and when available, validated siRNAs generated with highest potency and specificity in mind so you can focus on the highest data quality backed by the best in industry guarantee.Silencer Select siRNA libraries empower you with highly potent siRNAs to maximize discovery with even lessInvitrogen Silencer Select siRNAs are designed with the highest potency of available siRNAs, resulting in 100-fold more potent siRNAs and allowing gene knockdowns at the lowest concentrations. The highly potent Silencer Select siRNAs allow you to use even lower concentrations of siRNAs, which helps to minimize off target effects, reduce false negative data and maximize siRNA library output for additional screens and data validation. The Silencer Select Human Cancer Genome siRNA Library contains 0.25 nmol of each highly potent siRNA, sufficient for at least 500 transfections (at 5 nM siRNA, 100 μL transfection volume).
The Silencer Select siRNAs are designed using Thermo Fisher Scientific’s leading algorithms—along with our novel locked nucleic acid (LNA) chemical modifications, this helps to maximize the efficacy, specificity and potency required for RNAi functional screening against cancer gene targets.
  • Superior efficacy—Reduce off target effects by up to 90% due to latest design algorithms and LNA chemical modifications
  • Highest efficiency—Perform knockdown studies with 100-fold more potent siRNAs than currently available siRNAs
  • Dependable—Achieve faster discoveries via consistency of gene knockdown and reliability of phenotypic results
  • Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats
TRUSTED_SUSTAINABILITY
Spezifikation
Description 1596 total siRNAs targeting 532 genes plated onto 96-well plates (21 plates)
For research use only. Not for use in diagnostic procedures
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.