missing translation for 'onlineSavingsMsg'
Learn More

PeproTech EMAP II Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Product Code. 17844553
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17844553 25 μg 25µg
17823933 50 μg 50µg
17894563 2 x 500 μg 1mg
30054430 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17844553 Supplier PeproTech Supplier No. 500P172GBT25UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: discontinued. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human EMAP-II. Anti-Human EMAP-II-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hEMAP-II by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human EMAP-II (500-P172G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hEMAP-II. Western Blot: To detect hEMAP-II by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hEMAP-II is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P172GBT-1MG will be provided as 2 x 500 μg

The protein encoded by this gene is a cytokine that is specifically induced by apoptosis, and it is involved in the control of angiogenesis, inflammation, and wound healing. The release of this cytokine renders the tumor-associated vasculature sensitive to tumor necrosis factor. The precursor protein is identical to the p43 subunit, which is associated with the multi-tRNA synthetase complex, and it modulates aminoacylation activity of tRNA synthetase in normal cells. This protein is also involved in the stimulation of inflammatory responses after proteolytic cleavage in tumor cells. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene has been identified on chromosome 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMAP II
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Aimp1
Gene Accession No. Q12904
Gene Alias 9830137A06Rik; aaRS-interacting multifunctional protein 1; AIMP1; AIMP1/p43; AIMP1-PA; Aminoacyl tRNA synthase complex-interacting multifunctional protein 1; aminoacyl tRNA synthetase complex interacting multifunctional protein 1; aminoacyl tRNA synthetase complex-interacting multifunctional protein 1; aminoacyl-tRNA synthetase-interacting multifunctional protein 1; ARS inducible multifunctional protein 1; ARS-interacting multifunctional protein 1; CG8235; CG8235-PA; Dmel\CG8235; Dmel_CG8235; EMAP2; EMAP-2; EMAPII; EMAP-II; endothelial monocyte activating polypeptide 2; endothelial monocyte-activating polypeptide 2; Endothelial monocyte-activating polypeptide II; endothelial-monocyte activating polypeptide II; HLD3; mca1; multisynthase complex auxiliary component p43; multisynthetase complex auxiliary component p43; p43; SCYE1; Small inducible cytokine subfamily E member 1; small inducible cytokine subfamily E, member 1; small inducible cytokine subfamily E, member 1 (endothelial monocyte-activating)
Gene Symbols Aimp1
Host Species Goat
Immunogen E.coli-derived Recombinant Human EMAP-II.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9255
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.