missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ 96-Well Assay Plates for Cell-Based Assays

Product Code. 10256102 Shop All Merck Products
Click to view available options
Pore Size:
3 μm
5 μm
8 μm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10256102

Brand: Merck MAMIC3S10

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed

Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Polycarbonate
Array 8 x 12
Pore Size 3 μm
Color Clear
Material Styrene
No. of Wells 96
Sterility Sterile
Surface Treatment Tissue Culture
Filtration Area 0.28 cm2
For Use With (Application) Migration, invasion and chemotaxis
Quantity 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.