missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ 96-Well Assay Plates for Cell-Based Assays

Product Code. 10755614 Shop All Merck Products
Change view
Click to view available options
Pore Size:
3 μm
5 μm
8 μm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
10755614 8 μm Pack of 10
10256102 3 μm Pack of 10
10486892 5 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10755614 Supplier Merck Supplier No. MAMIC8S10
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed

Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Polycarbonate
Array 8 x 12
Pore Size 8 μm
Color Clear
Product Type MultiScreen-MIC Plate
Material Styrene
No. of Wells 96
Sterility Sterile
Surface Treatment Tissue Culture
Filtration Area 0.28 cm2
For Use With (Application) Migration, invasion and chemotaxis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.