missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ 96-Well Assay Plates for Cell-Based Assays
Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed
1700.00 €
Specifications
Format | Standard |
---|---|
Membrane | Polycarbonate |
Array | 8 x 12 |
Color | Clear |
Material | Styrene |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10256102
|
Merck
MAMIC3S10 |
3 μm |
1700.00 €
Pack of 10 |
Estimated Shipment: 26-08-2024
Log in to see stock. |
|||||
10486892
|
Merck
MAMIC5S10 |
5 μm |
1700.00 €
Pack of 10 |
Estimated Shipment: 26-08-2024
Log in to see stock. |
|||||
10755614
|
Merck
MAMIC8S10 |
8 μm |
1700.00 €
Pack of 10 |
Estimated Shipment: 26-08-2024
Log in to see stock. |
|||||
Description
Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
Specifications
Standard | |
8 x 12 | |
Styrene | |
Sterile | |
Migration, invasion and chemotaxis |
Polycarbonate | |
Clear | |
96 | |
Tissue Culture |