missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Shop All Merck Products
Click to view available options
Color:
Clear
White
Material:
Acrylic
Polystyrene
Sterility:
Non-sterile
Sterile
Membrane:
Hydrophobic PVDF
Hydrophobic PVDF with Underdrain
Mixed Cellulose Esters
Unit Size:
Pack of 10
Pack of 50
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Specifications
| Format | Standard |
| Membrane | Hydrophobic PVDF |
| Array | 8 x 12 |
| Pore Size | 0.45 μm |
| Product Line | MultiScreen |
| Color | White |
| Product Type | MultiScreen IP |
| Material | Acrylic |
| No. of Wells | 96 |
| Sterility | Sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction