Learn More
Merck MultiScreen™ Column Loaders
No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly
Brand:  Merck MACL09625
Code : 73
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10760113
  • 950.00 € / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket
Description
Description
  • Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
Specifications
Specifications
Standard
Black
25 μL
Non-sterile
1/Pk.
8 x 12
Aluminum
96
Load 25μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography