missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ High-Volume Manifold Collar

Product Code. 15649913 Shop All Merck Products
Change view
Click to view available options
Material:
Nylon
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15649913 Nylon Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15649913 Supplier Merck Supplier No. MSVMHTSHV
  • ONLINE EXCLUSIVE PRICE
    690.00 €
    637.07 € / Pack of 1
    Save 52.93 €
    7% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with MultiScreen high-volume filter plates

  • Fits over High-Volume Collection Plates
  • Opaque nylon construction

TRUSTED_SUSTAINABILITY

Specifications

Color Opaque
Material Nylon
For Use With (Application) Vacuum filtration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.