Learn More
Merck MultiScreen™ Multiple Punch and Punch Tips
Product Code. 10156100 Shop All Merck Products
Change view
Click to view available options
Product Type:
Punch kit
Punch tip
Unit Size:
Pack of 10
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10156100 Punch tip Pack of 50
11546365 Punch kit Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10156100 Supplier Merck Supplier No. MADP19650
  • ONLINE EXCLUSIVE PRICE
    482.00 €
    445.07 € / Pack of 50
    Save 36.93 €
    7% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For rdionucleotide counting by transfer of individual filters to tubes
  • Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
  • Remove filters from wells separately, eliminating cross-contamination
  • Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
  • Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
TRUSTED_SUSTAINABILITY
Specifications
Product Type Punch tip
For Use With (Application) Radionucleotide counting by transfer of individual filters to tubes
Quantity 50/Cs.